missing translation for 'onlineSavingsMsg'
Learn More

CD24, Biotin, clone: eBioSN3 (SN3 A5-2H10), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-0247-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15239499

  • £43.71 / 25µg


Estimated Shipment date
from Supplier 26-06-2018
Add to basket

Description

Description

Description: The SN3 monoclonal antibody reacts with the human CD24 molecule, also known as Heat Stable Antigen (HSA). This 35-50kDa molecule is anchored in the plasma membrane via phosphatidylinositol and is expressed by erythrocytes, thymocytes, peripheral lymphocytes and myeloid lineage. It has been reported that P-selectin (CD62P) binds to CD24.Applications Reported: This eBioSN3 (SN3 A5-2H10) antibody has been reported for use in flow cytometric analysis.Applications Tested: This eBioSN3 (SN3 A5-2H10) antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD24
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P25063
CD24 antigen (small cell lung carcinoma cluster 4 antigen), CD24A, FLJ22950, FLJ43543, MGC75043
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
eBioSN3 (SN3 A5-2H10)
Biotin
CD24
Conjugated
CD24
IgG1, kappa
25μg
Store at 2-8°C. Do not freeze.
Primary
100133941
Videos

For Research Use Only.