missing translation for 'onlineSavingsMsg'
Learn More

CD235a (Glycophorin A), eFluor 450, clone: HIR2 (GA-R2), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 48-9987-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15549856

  • £82.67 / 25 tests


Estimated Shipment date
from Supplier 29-05-2018
Add to basket

Description

Description

Description: The HIR2 monoclonal antibody reacts with human glycophorin A, a sialoglycoprotein expressed by erythroid precursors and mature circulating red cells.Applications Reported: This HIR2 (also GA-R2) antibody has been reported for use in flow cytometric analysis.Applications Tested: This HIR2 (also GA-R2) antibody has been pre-titrated and tested by flow cytometric analysis of diluted normal human peripheral blood cells. Binding of this antibody to red cells at high antibody concentration causes cell agglutination. This can be used at 5μL (0.03μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor® 450 is an alternative to Pacific Blue®. eFluor® 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD235a (Glycophorin A)
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P02724
Glycophorin A
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
HIR2 (also GA-R2)
eFluor 450
GYPA
Conjugated
GYPA
IgG2b, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
2993
Videos

For Research Use Only.