missing translation for 'onlineSavingsMsg'
Learn More

CD235a (Glycophorin A), clone: HIR2 (GA-R2), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-9987-80

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15258317

  • £30.74 / 25µg


Estimated Shipment
08-06-2018
Add to basket

Description

Description

Description: The HIR2 monoclonal antibody reacts with human glycophorin A, a sialoglycoprotein expressed by erythroid precursors and mature circulating red cells.Applications Reported: HIR2 can be used in flow cytometric analysis.Applications Tested: The HIR2 (also GA-R2) antibody has been tested by flow cytometric analysis of normal human peripheral blood cells and cell agglutination assay and can be used at approximately 0.1μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD235a (Glycophorin A)
Monoclonal
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
P02724
Glycophorin A
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
HIR2 (also GA-R2)
Unconjugated
GYPA
Purified
GYPA
IgG2b, kappa
25μg
Store at 2-8°C.
Primary
2993
Videos

For Research Use Only.