missing translation for 'onlineSavingsMsg'
Learn More

CD235a (Glycophorin A), Biotin, clone: HIR2 (GA-R2), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-9987-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15350150

  • £49.78 / 25µg


EU Stock 2
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The HIR2 monoclonal antibody reacts with human glycophorin A, a sialoglycoprotein expressed by erythroid precursors and mature circulating red cells.Applications Reported: This HIR2 (also GA-R2) antibody has been reported for use in flow cytometric analysis.Applications Tested: This HIR2 (also GA-R2) antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. Binding of this antibody to red cells at high antibody concentration causes cell agglutination. This can be used at less than or equal to 0.125μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD235a (Glycophorin A)
HIR2 (also GA-R2)
Biotin
Conjugated
IgG2b
RUO
Primary
Human
Flow Cytometry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Murine
25μg
Store at 2-8°C. Do not freeze.
Monoclonal
Videos