Learn More
CD23 Monoclonal Antibody (Tu1), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MHCD2304
Code : 50
Additional Details : Weight : 0.50000kg
Description
R-phycoerythrin (PE) is a stable and highly soluble phycobiliprotein which provides maximal absorbance and fluorescence without susceptibility to internal or external fluorescence quenching, thus providing an exceptional quantum yields and molar extinction coefficients.
CD23 Monoclonal antibody specifically detects CD23 in Human samples. It is validated for Flow Cytometry.Specifications
CD23 | |
Monoclonal | |
PE | |
FCER2 | |
BLAST-2; CD23; CD23 antigen; CD23A; CLEC4J; C-type lectin domain family 4 member J; C-type lectin domain family 4, member J; Fc epsilon receptor II; FC epsilon RII; Fc fragment of IgE receptor II; Fc fragment of IgE, low affinity II, receptor for (CD23); Fc receptor alpha multiple ligand receptor; Fc receptor, IgE, low affinity II, alpha polypeptide; Fcalpha muR; Fcalpha RII; Fce2; fc-epsilon-RII; FCER2; Fcer2a; FceRII; IGEBF; Immunoglobulin E-binding factor; immunoglobulin epsilon-chain; low affinity immunoglobulin epsilon Fc receptor; Low affinity immunoglobulin epsilon Fc receptor membrane-bound form; Low affinity immunoglobulin epsilon Fc receptor soluble form; low-affinity IgE receptor; Ly-42; lymphocyte IgE receptor | |
Mouse | |
purified | |
ASR, ASR/IVD | |
2208 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
Tu1, Tü1 | |
PBS with BSA, sucrose and 0.1% sodium azide | |
P06734 | |
FCER2 | |
Human CD23 | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG3 |