missing translation for 'onlineSavingsMsg'
Learn More

CD23, PE, clone: EBVCS2, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-0238-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15536736

  • £56.31 / 25 tests


Estimated Shipment date
from Supplier 27-03-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The EBVCS2 monoclonal antibody reacts with human CD23, a 45kDa type II transmembrane glycoprotein. CD23 is expressed on mature B cells, mantle zone B cells, follicular dendritic cells and at low levels on T, NK, langerhans cells and platelets. Expression of CD23 is upregulated upon B cell activation, and soluble forms of the antigen have been reported to be biologically active. CD23 is a low affinity receptor for IgE and is thought to play a role in the regulation of IgE response and B cell activation. CD21 and the alpha subunit of CD11b and CD11c bind to CD23.Applications Reported: The EBVCS2 antibody has been reported for use in flow cytometric analysis.Applications Tested: This EBVCS2 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD23
EBVCS2
PE
Conjugated
Murine
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (0.125μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Low Affinity IgE Receptor, FceRII, FcER2, IGEBF
IgG1
RUO
Primary
Human
Videos