missing translation for 'onlineSavingsMsg'
Learn More

Mouse CD229/SLAMF3 Antibody, R&D Systems™

Goat Polyclonal Antibody

POA

Specifications

Antigen CD229/SLAMF3/Lymphocyte Antigen 9
Concentration LYOPH
Dilution Western Blot 0.1 ug/mL
Applications Western Blot
Classification Polyclonal
View More Specs

Description

Description

CD229/SLAMF3/Lymphocyte Antigen 9 Polyclonal specifically detects CD229/SLAMF3/Lymphocyte Antigen 9 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

CD229/SLAMF3/Lymphocyte Antigen 9
Western Blot 0.1 ug/mL
Polyclonal
Goat
Mouse
CD229, CD229 antigen, cell-surface molecule Ly-9, hly9, Ly9, lymphocyte antigen 9Cell surface molecule Ly-9, mLY9, SLAMF3, T-lymphocyte surface antigen Ly-9
LY9
IgG
Affinity Purified
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
LYOPH
Western Blot
Unconjugated
RUO
AAH95921
4063
Mouse myeloma cell line NS0-derived recombinant mouse CD229/SLAMF3 Lys48-Phe454 Accession # AAH95921
Primary
Reconstitute at 0.2 mg/mL in sterile PBS.
Detects mouse CD229/SLAMF3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 20% cross-reactivity with recombinant human CD229 is observed.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only