Learn More
CD22 Monoclonal Antibody (RFB-4), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MHCD2204
Code : 50
Additional Details : Weight : 0.50000kg
Description
R-phycoerythrin (PE) is a stable and highly soluble phycobiliprotein which provides maximal absorbance and fluorescence without susceptibility to internal or external fluorescence quenching, thus providing an exceptional quantum yields and molar extinction coefficients.
CD22 Monoclonal antibody specifically detects CD22 in Human samples. It is validated for Flow Cytometry.Specifications
CD22 | |
Monoclonal | |
PE | |
CD22 | |
A530093D23; B-cell receptor CD22; BL-CAM; B-lymphocyte cell adhesion molecule; B-lymphocyte cell adhesion molecule (BL-CAM); Cd22; CD22 antigen; CD22 molecule; FLJ22814; Lectin 2; Leu-14; Lyb8; Lyb-8; MGC130020; sialic acid binding Ig-like lectin 2; sialic acid-binding Ig-like lectin 2; Sialic acid-binding Ig-like lectin 2 (Siglec-2); Siglec2; Siglec-2; T-cell surface antigen Leu-14 | |
Mouse | |
purified | |
ASR | |
933 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
RFB-4 | |
PBS with BSA, sucrose and 0.1% sodium azide | |
P20273 | |
CD22 | |
Human CD22 | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |