missing translation for 'onlineSavingsMsg'
Learn More

CD22, APC, clone: S-HCL-1, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-4229-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15508096

  • £68.27 / 25 tests

Estimated Shipment date
from Supplier 25-04-2018
Add to basket



Description: The S-HCl-1 monoclonal antibody recognizes human CD22 (Siglec-2), which is a member of the siglec subgroup of the Ig superfamily. CD22 is a type I transmembrane glycoprotein composed of two polypeptide chains, CD22a and CD22β, of 130 and 140kDa respectively, produced by alternative splicing of the CD22 gene. CD22 is expressed at high levels on mature B cells and B cell lymphomas. The extracellular portion of CD22 contains seven Ig-like domains, some of which are capable of binding ligands with sialic acid moieties expressed on epithelial, endothelial, B and T cells. The intracellular portion of CD22 contains 6 tyrosine residues contained within immunotyrosine-based inhibitory motifs (ITIM) and immunotyrosine-based activation-like motifs, which are phosphorylated upon B-cell receptor engagement, which enables CD22 to participate in the positive and negative regulation of B-cell receptor signaling.Applications Reported: This S-HCL-1 antibody has been reported for use in flow cytometric analysis.Applications Tested: This S-HCL-1 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.


25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Flow Cytometry
5μL (0.125μg)/test
aqueous buffer, 0.09% sodium azide, contains stabilizer if necessary
siglec2, siglec 2
IgG2b, kappa

For Research Use Only.