CD218a (IL-18 Receptor alpha) Mouse anti-Human, PE, Clone: H44, eBioscience™

Mouse Monoclonal Antibody

Overview
Brand: Affymetrix eBioscience

Manufacturer Part Number: 12-7183-41

Code: NEW

Additional Details:
Additional Details: Weight: 0.23750kg



Disclaimers: For Research Use Only.

Product Code. 15547106

Quantity Price
1 £ 91.11 / 25 tests
Estimated Shipment date
from Supplier 01-06-2017
Add to basket


Description and Specification

Specification

Antigen CD218a (IL-18 Receptor alpha)
Applications Flow Cytometry
Clone H44
Concentration 5μL (0.25μg)/test
Conjugate PE
Format Conjugated
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Gene Alias IL-1Rrp
Host Species Murine
Isotype IgG1
Quantity 25 Tests
Regulatory Status RUO
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Target Species Human

The H44 antibody reacts with the interleukin-18 receptor alpha chain, also known as IL-1Rrp, an IL-1R family member. IL-18R alpha is expressed on Th1 cells, NK cell subset, neutrophils and IL-12-stimulated tonsillar B cells. By itself, IL-18R alpha binds IL-18 with low affinity and when complexed with IL-18R beta chain binds IL-18 with high affinity. The proinflammatory cytokine IL-18 and its receptor complex play an important role in the innate and autoimmune responses.