missing translation for 'onlineSavingsMsg'
Learn More

CD218a (IL-18Ra), FITC, clone: H44, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-7183-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15556596

  • £87.94 / 25 tests


EU Stock 1
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The H44 antibody reacts with the interleukin-18 receptor alpha chain, also known as IL-1Rrp, an IL-1R family member. IL-18R alpha is expressed on Th1 cells, NK cell subset, neutrophils and IL-12-stimulated tonsillar B cells. By itself, IL-18R alpha binds IL-18 with low affinity and when complexed with IL-18R beta chain binds IL-18 with high affinity. The proinflammatory cytokine IL-18 and its receptor complex play an important role in the innate and autoimmune responses.Applications Reported: H44 has been reported for use in flow cytometric analysis.Applications Tested: This H44 antibody has been pre-titrated and tested by flow cytometric analysis on peripheral blood cells and Granulocytes. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD218a (IL-18 Receptor alpha)
H44
FITC
Conjugated
Murine
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (0.25μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
IL-1Rrp
IgG1
RUO
Primary
Human
Videos