missing translation for 'onlineSavingsMsg'
Learn More

CD218a (IL-18Ra) Mouse anti-Human, APC, Clone: H44, eBioscience™

Mouse Monoclonal Antibody

£97.75 - £190.00

Specifications

Antigen CD218a (IL-18Ra)
Clone H44
Target Species Human
Conjugate APC
Isotype IgG1, kappa, kappa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15598186
View Documents
Affymetrix eBioscience
17-7183-41
25 Tests £97.75
25 tests View alternative products
Save Up to 
»
Estimated Shipment
02-11-2020
Please to purchase this item. Need a web account? Register with us today!
15508196
View Documents
Affymetrix eBioscience
17-7183-42
100 Tests £190.00
100 tests View alternative products
Save Up to 
»
Estimated Shipment
02-11-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

IL18R1 is a cytokine receptor that belongs to the interleukin 1 receptor family. This receptor specifically binds interleukin 18 (IL18), and is essential for IL18 mediated signal transduction. IFN-alpha and IL12 are reported to induce the expression of this receptor in NK and T cells. The gene along with four other members of the interleukin 1 receptor family, including IL1R2, IL1R1, ILRL2 (IL-1Rrp2), and IL1RL1 (T1/ST2), form a gene cluster on chromosome 2q.
Specifications

Specifications

CD218a (IL-18Ra)
Human
IgG1, kappa, kappa
Monoclonal
Liquid
Antibody
Store at 2-8°C. Do not freeze. Light-sensitive material.
H44
APC
Flow Cytometry
Primary
Monoclonal
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
RUO

For Research Use Only.