missing translation for 'onlineSavingsMsg'
Learn More

CD218a (IL-18Ra) Mouse anti-Human, APC, Clone: H44, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-7183-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15598186

  • £97.75 / 25 tests

Estimated Shipment
02-10-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: The H44 antibody reacts with the interleukin-18 receptor alpha chain, also known as IL-1Rrp, an IL-1R family member. IL-18R alpha is expressed on Th1 cells, NK cell subset, neutrophils and IL-12-stimulated tonsillar B cells. By itself, IL-18R alpha binds IL-18 with low affinity and when complexed with IL-18R beta chain binds IL-18 with high affinity. The proinflammatory cytokine IL-18 and its receptor complex play an important role in the innate and autoimmune responses.Applications Reported: This H44 antibody has been reported for use in flow cytometric analysis.Applications Tested: This H44 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.It is recommended to stain whole blood before RBC lysis.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD218a (IL-18Ra)
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q13478
Interleukin-18
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
H44
APC
IL18R1
Liquid
IL18R1
IgG1, kappa, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
8809
Videos
Documents

For Research Use Only.