missing translation for 'onlineSavingsMsg'
Learn More

CD217 (IL-17Ra) Mouse anti-Human, PE, Clone: J10MBS, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-7517-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15567126

  • £92.25 / 25 tests

Estimated Shipment
10-08-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Interleukin 17A (IL-17A) is a proinflammatory cytokine secreted by activated T-lymphocytes. It is a potent inducer of the maturation of CD34-positive hematopoietic precursors into neutrophils. IL-17RA is a ubiquitous type I membrane glycoprotein that binds with low affinity to IL-17A and IL-17F. IL-17A and its receptor play a pathogenic role in many inflammatory and autoimmune diseases such as rheumatoid arthritis. Defects in IL-17RA are the cause of familial candidiasis type 5 (CANDF5) which is a rare disorder with altered immune responses and impaired clearance of fungal infections.
Specifications

Specifications

CD217 (IL-17Ra)
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q96F46
interleukin 17 receptor A; AW538159; Cdw217; IL17r; VDw217; interleukin 17 receptor
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
J10MBS
PE
IL17RA
Liquid
IL17RA
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
23765
Videos
Documents

For Research Use Only.