missing translation for 'onlineSavingsMsg'
Learn More

CD217 (IL-17Ra) Mouse anti-Human, PE, Clone: J10MBS, eBioscience™

Mouse Monoclonal Antibody

£92.25 - £198.00

Specifications

Antigen CD217 (IL-17Ra)
Clone J10MBS
Target Species Human
Conjugate PE
Isotype IgG1, kappa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15567126
View Documents
Affymetrix eBioscience
12-7517-41
25 Tests £92.25
25 tests View alternative products
Save Up to 
»
Estimated Shipment
26-11-2020
Please to purchase this item. Need a web account? Register with us today!
15577126
View Documents
Affymetrix eBioscience
12-7517-42
100 Tests £198.00
100 tests View alternative products
Save Up to 
»
Estimated Shipment
26-11-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Interleukin 17A (IL-17A) is a proinflammatory cytokine secreted by activated T-lymphocytes. It is a potent inducer of the maturation of CD34-positive hematopoietic precursors into neutrophils. IL-17RA is a ubiquitous type I membrane glycoprotein that binds with low affinity to IL-17A and IL-17F. IL-17A and its receptor play a pathogenic role in many inflammatory and autoimmune diseases such as rheumatoid arthritis. Defects in IL-17RA are the cause of familial candidiasis type 5 (CANDF5) which is a rare disorder with altered immune responses and impaired clearance of fungal infections.
Specifications

Specifications

CD217 (IL-17Ra)
Human
IgG1, kappa
Monoclonal
Liquid
Antibody
4° C, store in dark, DO NOT FREEZE!
J10MBS
PE
Flow Cytometry
Primary
Monoclonal
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
RUO

For Research Use Only.