missing translation for 'onlineSavingsMsg'
Learn More

CD21, PE, clone: HB5, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-0219-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15506736

  • £61.93 / 25 tests


Estimated Shipment date
from Supplier 03-07-2018
Add to basket

Description

Description

Description: The HB5 monoclonal antibody reacts with human CD21, an approximately 145kDa type I transmembrane protein. CD21 is expressed by mature B cells, folliculate dendritic cells and a subset of epithelial cells and is a receptor for complement component C3d and Epstein-Barr virus (EBV). CD21, in association with CD19 and CD81, participates in the multimolecular complex with BCR and is involved in B cell activation.Applications Reported: This HB5 antibody has been reported for use in flow cytometric analysis.Applications Tested: This HB5 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD21
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P20023
EBV receptor, complement C3d receptor, complement receptor type 2, epstein-Barr virus receptor
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
HB5
PE
CR2
Conjugated
CR2
IgG2a, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
1380
Videos

For Research Use Only.