missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD21 (Mature B-Cell & Follicular Dendritic Cell Marker) Mouse, Clone: CR2/1953, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 1380-MSM3-P0
Additional Details : Weight : 0.01000kg
Specifications
CD21 (Mature B-Cell & Follicular Dendritic Cell Marker) | |
Cell Surface | |
CR2/1953 | |
Unconjugated | |
Mouse | |
140kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
1380 | |
IgG1 κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant fragment (apprx. aa 44-196) of human CR2 (CD21) protein | |
WEHI-231 cells and Tonsil | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |