Learn More
CD21/CD35 Monoclonal Antibody (eBio8D9 (8D9)), PE-Cyanine7, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£67.25 - £262.00
Specifications
Antigen | CD21/CD35 |
---|---|
Clone | eBio8D9 (8D9) |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15380900
|
Affymetrix eBioscience
25-0211-82 |
100 μg |
£262.00
100µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15370900
|
Affymetrix eBioscience
25-0211-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The monoclonal antibody eBio8D9 reacts with an eptiope shared by mouse CD21 (CR2) and CD35 (CR1). CD21 and CD35 are alternatively spliced transcripts from the Cr2 gene, which produce cell-surface proteins of 145 and 190 kDa, respectively. CD21 and CD35 are expressed by mature B cells, but not on thymocytes, peripheral T cells, erythrocytes or platelets. Furthermore, there is some evidence which demonstrates their expression on macrophages. CD21 is a receptor for the complement component C3d and Epstein-Barr virus (EBV). In association with CD19 and CD81, CD21 also participates in B-cell activation through the B cell receptor. Cr2-deficient mice display impaired inflammatory and humoral immune responses in vivo. The anti-mouse CD21/35 monoclonal antibody clones eBio4E3 and eBio8D9 do not cross-block each other, suggesting that they bind to different epitopes. Applications Reported: This eBio8D9 (8D9) antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBio8D9 (8D9) antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
CD21/CD35 Monoclonal antibody specifically detects CD21/CD35 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD21/CD35 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
P19070 | |
Cr2 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Cr2 |
eBio8D9 (8D9) | |
Flow Cytometry | |
PE-Cyanine7 | |
Rat | |
Mouse | |
12902 | |
IgG2a λ | |
Affinity chromatography | |
Antibody |
For Research Use Only.