missing translation for 'onlineSavingsMsg'
Learn More

CD21/CD35, eFluor 450, clone: eBio4E3 (4E3), eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 48-0212-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15311700

  • £81.20 / 25µg


EU Stock 2
Add to basket

Description

Description

Description: The monoclonal antibody eBio4E3 reacts with an eptiope shared by mouse CD21 (CR2) and CD35 (CR1). CD21 and CD35 are alternatively spliced transcripts from the Cr2 gene, which result in cell-surface proteins of 145 and 190 kDa, respectively. CD21 and CD35 are expressed by mature B cells, but not on thymocytes, peripheral T cells, erythrocytes or platelets. Furthermore, there is little evidence which demonstrates their expression on macrophages. CD21 is a receptor for complement component C3d and Epstein-Barr virus (EBV), and in association with CD19 and CD81, CD21 also participates in B-cell activation through the B cell receptor. Cr2 deficient mice display impaired inflammatory and humoral immune responses in vivo. The anti-mouse CD21/35 monoclonal antibody clones eBio4E3 and eBio8D9 do not cross-block each other suggesting that they bind to different epitopes.Applications Reported: This eBio4E3 (4E3) antibody has been reported for use in flow cytometric analysis.Applications Tested: This eBio4E3 (4E3) antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.eFluor® 450 is an alternative to Pacific Blue®. eFluor® 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD21/CD35
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P19070
EBV receptor, complement C3d receptor, complement receptor type 2, epstein-Barr virus receptor, C3-binding protein, C3b/C4b receptor, CD35 antigen, Knops blood group antigen, complement component receptor 1, complement receptor type 1
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
eBio4E3 (4E3)
eFluor 450
Cr2
Conjugated
Cr2
IgG2a, lambda
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
12902
Videos

For Research Use Only.