missing translation for 'onlineSavingsMsg'
Learn More

CD209a Monoclonal Antibody (MMD3), Alexa Fluor™ 488, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 53-2094-80

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15540207

  • £166.00 / 25µg
Estimated Shipment: 22-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: This MMD3 monoclonal antibody recognizes mouse CD209a, which is also known as DC-SIGN. CD209a is a type II transmembrane C-type lectin expressed on a subset of dendritic cells, including some CD4+, CD8- and plasmacytoid pre-dendritic cells. Studies indicate that CD209a expression can vary according to the activation state of the host. Moreover, CD209a is down-regulated in spleen-derived dendritic cell cultures supplemented with GM-CSF. CD209a is involved in mediating the innate immune response by binding microbial carbohydrates. Cross-blocking studies suggest that MMD3 recognizes a different a different epitope from LWC06 and 5H10. Applications Reported: This MMD3 antibody has been reported for use in immunohistochemical staining of frozen tissue sections, microscopy, immunohistochemical staining, and immunocytochemistry. Applications Tested: This MMD3 antibody has been tested by immunohistochemistry on frozen 5-week-old mouse thymus and can be used at less than or equal to 20 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.

CD209a Monoclonal antibody specifically detects CD209a in Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen).
Specifications

Specifications

CD209a
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q91ZX1
Cd209a
Affinity chromatography
RUO
170786
4° C, store in dark, DO NOT FREEZE!
Liquid
Immunocytochemistry, Immunohistochemistry (Frozen)
MMD3
Alexa Fluor 488
Cd209a
CD209; CD209 antigen; CD209 antigen-like protein A; CD209 molecule; Cd209a; CD209a antigen; CD209a molecule; Cd209d; CD209d antigen; CDSIGN; Cire; CLEC4L; Clec4m; C-type lectin domain family 4 member L; C-type lectin domain family 4, member L; C-type lectin domain family 4, member M; Dcsign; DC-SIGN; DC-SIGN1; Dc-signr; DC-SIGN-related protein; Dendritic cell-specific ICAM-3-grabbing non-integrin; dendritic cell-specific ICAM-3-grabbing non-integrin 1; dendritic cell-specific intercellular adhesion molecule-3-grabbing non-integrin; dendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbing non-integrin; HIV gpl20-binding protein; MGC129965; MGC130443; RGD1561104; SIGN-R1; SIGNR5
Mouse
25 μg
Primary
Mouse
Antibody
IgG2c κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.