Learn More
CD209 (DC-SIGN) Rat anti-Mouse, Clone: 5H10, eBioscience™
Rat Monoclonal Antibody
Brand:  Affymetrix eBioscience 14-2091-80
UNSPSC : 12352200
Code : Z2
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15267307
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Disclaimers:
For Research Use Only.
Specifications
Description
The 5H10 antibody was generated by immunization with a peptide derived from the extracellular region of mouse CIRE/DC-SIGN (CD209). CIRE/DC-SIGN was identified by its expression on CD8α- dendritic cells and plasmacytoid predendritic cells, and is the closest homologue of human DC-SIGN. Human DC-SIGN was originally identified in human placenta for its ability to bind the HIV envelope protein gp120 in a CD4-independent manner. CIRE/DC-SIGN is a 33kDa type II transmembrane C-type lectin protein. It contains a C-terminal, extracellular, Carbohydrate Recognition Domain (CRD) that is predicted to bind mannose and other carbohydrates in a Ca2+ dependent manner. It has been postulated that CIRE/DC-SIGN may play a role in T-dendritic cell interactions through binding with members of the ICAM family. CIRE/DC-SIGN is differentially expressed by sub-populations of dendritic cells and preliminary data suggest that its expression varies depending on the activation state of the host. CIRE/DC-SIGN is down-regulated in spleen-derived dendritic cell cultures supplemented with GM-CSF. While human DC-SIGN is predominantly expressed in dendritic cells, CIRE/DC-SIGN mRNA has also been detected in B cells. The 5H10 monoclonal antibody does not cross-react with the closely related SIGNR1, SIGNR2, SIGNR3 or SIGNR4.
Description & Specifications
Specifications
CD209 (DC-SIGN)
5H10
Unlabeled
Purified
Rat
25μg
Store at 2-8°C.
Monoclonal
Western Blot
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
DCSIGN, CIRE
IgG2a
RUO
Primary
Murine