missing translation for 'onlineSavingsMsg'
Learn More

CD209 (DC-SIGN), FITC, clone: eB-h209, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-2099-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15526516

  • £62.18 / 25 tests


Estimated Shipment date
from Supplier 26-06-2018
Add to basket

Description

Description

Description: The eB-h209 monoclonal antibody reacts with human CD209, also known as DC-SIGN, a 44kDa type II transmembrane protein. DC-SIGN contains a C-type lectin binding domain and binds ICAM-3, ICAM-2, and HIV virus. Human dendritic cells preferentially express DC-SIGN. It has been postulated that DC-SIGN serves as a receptor for capture, trafficking, and transmission of HIV to T cells and supports primary immune response. eB-h209 was developed against a C-terminal peptide of human DC-SIGN.Applications Reported: The eB-h209 antibody has been reported for use in flow cytometric analysis.Applications Tested: This eB-h209 antibody has been pre-titrated and tested by flow cytometric analysis of cultured human dendritic cells and peripheral blood cells. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD209 (DC-SIGN)
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q9NNX6
CD209, CDSIGN, CIRE, DC-SIGN, DC-SIGN1, Dcsign, MGC130443, SIGN-R1, SIGNR5, CD209a
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
eB-h209
FITC
CD209
Conjugated
CD209
IgG2a, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
30835
Videos

For Research Use Only.