missing translation for 'onlineSavingsMsg'
Learn More

CD209 (DC-SIGN), clone: eB-h209, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-2099-80

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15227317

  • £87.94 / 25µg


Call for latest availability
+44 (0)1509 555 500
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The eB-h209 monoclonal antibody reacts with human CD209, also known as DC-SIGN, a 44kDa type II transmembrane protein. DC-SIGN contains a C-type lectin binding domain and binds ICAM-3, ICAM-2, and HIV virus. Human dendritic cells preferentially express DC-SIGN. It has been postulated that DC-SIGN serves as a receptor for capture, trafficking, and transmission of HIV to T cells and supports primary immune response. eB-h209 was developed against a C-terminal peptide of human DC-SIGN.Applications Reported: The eB-h209 antibody has been reported for use in flow cytometric analysis, and immunoprecipitation.Applications Tested: The eB-h209 antibody has been tested by flow cytometric analysis of cultured human dendritic cells and peripheral blood cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD209 (DC-SIGN)
eB-h209
Unconjugated
Purified
Rat
25μg
Store at 2-8°C.
Monoclonal
Immunoprecipitation
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Clec4L
IgG2a
RUO
Primary
Human
Videos