missing translation for 'onlineSavingsMsg'
Learn More

CD209 (DC-SIGN), Biotin, clone: eB-h209, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-2099-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15259779

  • £87.94 / 25µg


Estimated Shipment date
from Supplier 03-07-2018
Add to basket

Description

Description

Description: The eB-h209 monoclonal antibody reacts with human CD209, also known as DC-SIGN, a 44kDa type II transmembrane protein. DC-SIGN contains a C-type lectin binding domain and binds ICAM-3, ICAM-2, and HIV virus. Human dendritic cells preferentially express DC-SIGN. It has been postulated that DC-SIGN serves as a receptor for capture, trafficking, and transmission of HIV to T cells and supports primary immune response. eB-h209 was developed against a C-terminal peptide of human DC-SIGN.Applications Reported: The eB-h209 antibody has been reported for use in flow cytometric analysis.Applications Tested: The eB-h209 antibody has been tested by flow cytometric analysis of cultured human dendritic cells and peripheral blood cells This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD209 (DC-SIGN)
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q9NNX6
CD209, CDSIGN, CIRE, DC-SIGN, DC-SIGN1, Dcsign, MGC130443, SIGN-R1, SIGNR5, CD209a
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
eB-h209
Biotin
CD209
Conjugated
CD209
IgG2a, kappa
25μg
Store at 2-8°C. Do not freeze.
Primary
30835
Videos

For Research Use Only.