missing translation for 'onlineSavingsMsg'
Learn More

CD209 (DC-SIGN) Rat anti-Human, APC, Clone: eB-h209, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-2099-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15568056

  • £91.25 / 25 tests

Estimated Shipment
18-08-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a transmembrane receptor and is often referred to as DC-SIGN because of its expression on the surface of dendritic cells and macrophages. The encoded protein is involved in the innate immune system and recognizes numerous evolutionarily divergent pathogens ranging from parasites to viruses with a large impact on public health. The protein is organized into three distinct domains: an N-terminal transmembrane domain, a tandem-repeat neck domain and C-type lectin carbohydrate recognition domain. The extracellular region consisting of the C-type lectin and neck domains has a dual function as a pathogen recognition receptor and a cell adhesion receptor by binding carbohydrate ligands on the surface of microbes and endogenous cells. The neck region is important for homo-oligomerization which allows the receptor to bind multivalent ligands with high avidity. Variations in the number of 23 amino acid repeats in the neck domain of this protein are rare but have a significant impact on ligand binding ability. This gene is closely related in terms of both sequence and function to a neighboring gene (GeneID 10332; often referred to as L-SIGN). DC-SIGN and L-SIGN differ in their ligand-binding properties and distribution. Alternative splicing results in multiple variants.
Specifications

Specifications

CD209 (DC-SIGN)
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q9NNX6
CD209, CDSIGN, CIRE, DC-SIGN, DC-SIGN1, Dcsign, MGC130443, SIGN-R1, SIGNR5, CD209a
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
eB-h209
APC
CD209
Liquid
CD209
IgG2a, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
30835
Videos
Documents

For Research Use Only.