missing translation for 'onlineSavingsMsg'
Learn More

CD207 (Langerin), clone: eBioRMUL.2, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-2073-80

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15227307

  • £55.67 / 25µg


EU Stock 3
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: RMUL.2 is a monoclonal antibody that reacts with the predicted extracellular domain of mouse CD207, also known as Langerin. Mouse CD207 is expressed in a distinct subset of dendritic cells called Langerhans cells (LC). Mouse CD207 is a 48 kDa, C-type lectin and putative transmembrane protein that likely plays a role in antigen recognition and uptake. LC are located in the epidermis and upon activation reduce expression of CD207 and begin migrating through the dermis towards lymphatic vessels. Expression of mouse CD207 in LC has been correlated to the presence of Birbeck Granules. Furthermore, the formation of Birbeck Granules has been observed upon transfection of CD207 cDNA into fibroblast cell lines.Western Blotting with RMUL.2 reveals a 48kDa band in lysate from mouse ear epidermis as well as in lysates from cells transfected with mouse CD207 cDNA. Cross reactivity of this antibody with other proteins has not been determined. Preliminary data from mouse CD207 cDNA transfected cells suggests that RMUL.2 can be used for intracellular staining followed by flow cytometric analysis. We have observed in our transfectants the majority of protein is expressed intracellularly with a small percentage on the cell surface. In addition, RMUL.2 has also been tested by immunohistology on mouse ear epidermal sheets and shows specific staining.Applications Reported: This eBioRMUL.2 antibody has been reported for use in intracellular staining followed by flow cytometric analysis, immunoblotting (WB), and immunohistochemical staining. (Fluorochrome conjugated eBioRMUL.2 is recommended for use in intracellular flow cytometry.).Applications Tested: This eBioRMUL.2 antibody has been tested by immunoblotting. This can be used at less than or equal to 2 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD207 (Langerin)
eBioRMUL.2
Unconjugated
Purified
Rat
25μg
Store at 2-8°C.
Monoclonal
Western Blot
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Clec4K
IgG2a
RUO
Primary
Murine
Videos