missing translation for 'onlineSavingsMsg'
Learn More

CD205, PerCP-eFluor™ 710, clone: 205yekta, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 46-2051-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15371300

  • £83.62 / 25µg


Estimated Shipment
27-02-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The 205yekta antibody reacts with mouse CD205, also known as DEC-205 (dendritic and epithelial cells, 205 kDa). CD205 is an integral membrane glycoprotein involved in antigen uptake, trafficking and presentation that improves the induction of antigen-specific T cell immunity. CD205 is highly expressed by CD8+ dendritic cells (DCs) and also expressed at different levels by bone marrow Gr1+ cells, Langerhans cells, (BMDC) bone marrow derived DCs and thymic epithelial cells.CD205 has been shown to bind to PE-Cy5.5 antibody and streptavidin conjugates; therefore, this format should not be used on CD205+ dendritic cells.Applications Reported: This 205yekta antibody has been reported for use in flow cytometric analysis.Applications Tested: This 205yekta antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 0.03 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.PerCP-eFluor® 710 can be used in place of PE-Cy5, PE-Cy5.5 or PerCP-Cy5.5. PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm). Please make sure that your instrument is capable of detecting this fluorochrome. For a filter configuration, we recommend using the 685 LP dichroic mirror and 710/40 band pass filter, however the 695/40 band pass filter is an acceptable alternative. Our testing indicates that PerCP-eFluor® 710 conjugated antibodies are stable when stained samples are exposed to freshly prepared 2% formaldehyde overnight at 4°C, but please evaluate for alternative fixation protocols.Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD205
205yekta
PerCP-eFluor 710
Conjugated
Rat
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
DEC205, Ly75
IgG2a
RUO
Primary
Murine
Videos