missing translation for 'onlineSavingsMsg'
Learn More

CD203c, APC, clone: NP4D6, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-2039-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15518056

  • £81.61 / 25 tests


Estimated Shipment date
from Supplier 03-04-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: This NP4D6 monoclonal antibody reacts with human CD203c, which is also known as ectonucleotide pyrophosphatase/phosphodiesterase 3 (E-NPP3). ENPP family members catalyze the hydrolysis of pyrophosphate and phosphodiester bonds to nucleoside 5-monophosphates. A type II transmembrane protein, CD203c is expressed on basophils and mast cells. CD203c expression is upregulated on basophils activated by allergen or IgE crosslinking. Therefore, CD203c is used as an indicator of basophil activation in various IgE-mediated allergic responses.Applications Reported: This NP4D6 antibody has been reported for use in flow cytometric analysis.Applications Tested: This NP4D6 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD203c
NP4D6
APC
Conjugated
Murine
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (0.125μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
E-NPP3, PDNP3
IgG1
RUO
Primary
Human
Videos