missing translation for 'onlineSavingsMsg'
Learn More

CD202b (TIE2), PE, clone: TEK4, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-5987-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15289229

  • £80.57 / 50µg


EU Stock 3
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The TEK4 monoclonal antibody reacts with mouse Tie-2, also known as CD202. A member of the tyrosine kinase receptor family, Tie-2 is expressed on endothelial and a subset of hematopoietic cells and is believed to play a role in both angiogenesis and hematopoiesis during development of the mouse embryo. In fetal liver and adult bone marrow, Tie-2 is expressed by a subpopulation of hematopoietic progenitor cells characterized as Lineage markers^-, c-Kit^+, Sca1^+ cells. Long-term multilineage repopulating cells were detected in Tie-2^+, Lineage^-, c-Kit^+, Sca1^+ cells but not in Tie-2^-, Lineage^-, c-Kit^+, Sca1^+ cells.Applications Reported: The TEK4 antibody has been reported for use in flow cytometric analysis.Applications Tested: The TEK4 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 2 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD202b (TIE2)
TEK4
PE
Conjugated
Rat
50μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
TIE-2, TEK, CD202
IgG1
RUO
Primary
Murine
Videos