Learn More
CD201 (EPCR) Monoclonal Antibody (RCR-227), APC, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 17-2018-42
Code : NEW
Additional Details : Weight : 0.23750kg
Description
Description: The monoclonal antibody RCR-227 reacts with human CD201, which is also known as the endothelial protein C receptor (EPCR), a 46-kDa transmembrane protein related to class I MHC. CD201 was originally identified on vascular endothelium cells, predomitly larger blood vessels. It is also expressed on monocytes, lymphocytes, hematopoietic stem cells, neutrophils, and eosinophils. CD201 serves as a binding site for protein C, activated protein C (APC), and factor VII/VIIa. A soluble form has also been identified in the plasma. CD201 plays a role in controlling coagulation as disruption of its gene in mice caused placental thrombosis and early embryonic death. This cell surface molecule is also involved in the anti-inflammatory response. APC binding to CD201 enables PAR-1 activation that confers cytoprotective effects. In monocytes, APC binding decreases secretion of proinflammatory cytokines, chemokines, and growth factors. Finally, CD201 has been shown to inhibit lymphocyte migration. Applications Reported: This RCR-227 antibody has been reported for use in flow cytometric analysis. Applications Tested: This RCR-227 antibody has been pre-titrated and tested by flow cytometric analysis of HUVECs. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL.
CD201 (EPCR) Monoclonal antibody specifically detects CD201 (EPCR) in Human samples. It is validated for Flow Cytometry.Specifications
CD201 (EPCR) | |
Monoclonal | |
5 μL/Test | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
Q9UNN8 | |
Procr | |
Affinity chromatography | |
RUO | |
10544 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
RCR-227 | |
APC | |
Procr | |
Activated protein C receptor; AI325044; APC receptor; bA42O4.2; CCCA; CCD41; CD antigen CD201; CD201; CD201 antigen; cell cycle, centrosome-associated protein; centrocyclin; Centrosomal protein CCD41; endothelial cell protein C receptor; Endothelial protein C receptor; Epcr; PROCR; protein C receptor; protein C receptor, endothelial | |
Rat | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
For Research Use Only.