missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD200R1L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA549163
Additional Details : Weight : 0.01000kg
Description
CD200R1L Polyclonal antibody specifically detects CD200R1L in Human samples. It is validated for Western Blot.Specifications
CD200R1L | |
Polyclonal | |
Unconjugated | |
CD200R1L | |
CD200 cell surface glycoprotein receptor 2; CD200 cell surface glycoprotein receptor-like 2; CD200 cell surface glycoprotein receptor-like a; CD200 receptor 1 like; CD200 receptor 1-like; CD200 receptor 2; CD200 receptor-like 2; CD200R1L; CD200R2; CD200RLa; Cell surface glycoprotein CD200 receptor 1-like; cell surface glycoprotein CD200 receptor 2; Cell surface glycoprotein OX2 receptor 2; huCD200R2 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
344807 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 | |
PBS with 0.09% sodium azide | |
Q6Q8B3 | |
CD200R1L | |
KLH conjugated synthetic peptide between 63-92 amino acids from the N-terminal region of human CD200R1L. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |