missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD20 Mouse, FITC, Clone: 2H7, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio QAB31-F-500Tests
Additional Details : Weight : 0.01000kg
Specifications
CD20 | |
Monoclonal | |
5μL/Test | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
MS4A1 | |
Protein A/G Purified | |
RUO | |
931 | |
IgG2b κ |
Flow Cytometry | |
2H7 | |
FITC | |
NP_690605 | |
Mouse | |
500 Tests | |
Primary | |
Store at 2-4°C. |