missing translation for 'onlineSavingsMsg'
Learn More

CD20, eVolve 655, clone: 2H7, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 86-0209-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15580827

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Description: The 2H7 monoclonal antibody reacts with human CD20, a 33-36kDa transmembrane protein. CD20 is expressed by developing B cells as well as mature B cells but not plasma cells. CD20 has been detected at low levels on a small subset of mature T cells. It is suggested that CD20 plays a role in B-cell activation.Applications Reported: This 2H7 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 2H7 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blod cells. This can be used at 5 μL per test. A test is defined as the amount of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. For best results when using antibodies conjugated to eVolve™, we recommend a quick spin of the vial before use and avoid touching the bottom of the vial with the pipet tip.Laser/Filter Recommendation: eVolve™ 655 can be optimally excited with laser lines from 325-405 nm, and emits near 655 nm. We recommend using the violet (405 nm) laser line for excitation and a 630LP with a 660/40 bandpass filter, or equivalent, for detection.Fixation Recommendation: Antibodies conjugated to eVolve™ are compatible with eBioscience's fixation and permeabilization buffers.Excitation: 350-635 nm; Emission: 657 nm; Laser: Ultraviolet Laser, Violet LaserStorage and handling: For best results when using antibodies conjugated to eVolve™, we recommend a quick spin of the vial before use and avoid touching the bottom of the vial with the pipet tip.
Specifications

Specifications

CD20
Monoclonal
5μl/Test
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
P11836
B1, Bp35, CD20, LEU-16, MGC3969, MS4A1
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
2H7
eVolve 655
MS4A1
Conjugated
MS4A1
IgG2b, kappa
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material. This product should be used within 6 months from the date of receipt.
Primary
931
Videos
Documents

For Research Use Only.