Learn More
CD20 Monoclonal Antibody (2H7), APC-eFluor™ 780, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£98.25 - £232.00
Specifications
Antigen | CD20 |
---|---|
Clone | 2H7 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | APC-eFluor 780 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15569526
|
Affymetrix eBioscience
47-0209-42 |
100 Tests |
£232.00
100 tests |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15559526
|
Affymetrix eBioscience
47-0209-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The 2H7 monoclonal antibody reacts with human CD20, a 33-36 kDa transmembrane protein. CD20 is expressed by developing B cells as well as mature B cells but not plasma cells. CD20 has been detected at low levels on a small subset of mature T cells. It is suggested that CD20 plays a role in B-cell activation. Applications Reported: This 2H7 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 2H7 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blod cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. APC-eFluor 780 emits at 780 nm and is excited with the Red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Light sensitivity: This tandem is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation.
CD20 Monoclonal antibody specifically detects CD20 in Human samples. It is validated for Flow Cytometry.Specifications
CD20 | |
Flow Cytometry | |
APC-eFluor 780 | |
Mouse | |
Human | |
P11836 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |
2H7 | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
931 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
For Research Use Only.