missing translation for 'onlineSavingsMsg'
Learn More

CD20, APC, clone: 2H7, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-0209-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15507896

  • £75.06 / 25 tests


EU Stock 2
Add to basket

Description

Description

Description: The 2H7 monoclonal antibody reacts with human CD20, a 33-36kDa transmembrane protein. CD20 is expressed by developing B cells as well as mature B cells but not plasma cells. CD20 has been detected at low levels on a small subset of mature T cells. It is suggested that CD20 plays a role in B-cell activation.Applications Reported: This 2H7 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 2H7 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD20
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P11836
B1, Bp35, CD20, LEU-16, MGC3969, MS4A1
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
2H7
APC
MS4A1
Conjugated
MS4A1
IgG2b, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
931
Videos

For Research Use Only.