missing translation for 'onlineSavingsMsg'
Learn More

CD2, PE, clone: RM2-5, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-0021-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15278409

  • £74.54 / 50µg


Estimated Shipment date
from Supplier 03-07-2018
Add to basket

Description

Description

Description: The RM2-5 monoclonal antibody reacts with the mouse CD2 molecule, an approximately 50-55kDa cell surface receptor expressed by all mouse lymphocytes. Expression of the CD2 antigen in the mouse differs from that of the human in that B lineage cells in the mouse from the pre-B cell stage to mature B cells express CD2. CD2 is a ligand for CD48 in the mouse and is involved in adhesion and activation of T cells.Applications Reported: The RM2-5 antibody has been reported for use in flow cytometric analysis.Applications Tested: This RM2-5 antibody has been tested by flow cytometric analysis of mouse splenocytes and bone marrow cells. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD2
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P08920
CD2 antigen, sheep red blood cell receptor, LFA-3 receptor, T-cell surface antigen CD2, T-cell surface antigen T11/Leu-5, erythrocyte receptor, lymphocyte-function antigen-2, rosette receptor
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
RM2-5
PE
Cd2
Conjugated
Cd2
IgG2b, lambda
50μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
12481
Videos

For Research Use Only.