missing translation for 'onlineSavingsMsg'
Learn More

CD2 Mouse, FITC, Clone: RPA-2.10, enQuireBio™

Mouse Monoclonal Antibody

£109.00 - £985.00

Specifications

Antigen CD2
Clone RPA-2.10
Concentration 5μL/Test
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15961458
View Documents Promotion Details
EnquireBio
QAB1-F-25TESTS
25 Tests
£109.00
25 tests
View alternative products
Save Up to 
»
Estimated Shipment: 04-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15951458
View Documents Promotion Details
EnquireBio
QAB1-F-100TESTS
100 Tests
£219.00
100 tests
View alternative products
Save Up to 
»
Estimated Shipment: 04-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15971458
View Documents Promotion Details
EnquireBio
QAB1-F-500TESTS
500 Tests
£985.00
500 tests
View alternative products
Save Up to 
»
Estimated Shipment: 04-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

CD2
5μL/Test
Monoclonal
Mouse
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
914
IgG1 κ
Protein A/G Purified
RPA-2.10
Flow Cytometry
FITC
RUO
NP_001758
CD2
Primary
Store at 2-4°C.
Videos
SDS
Documents

Documents

Product Certifications