missing translation for 'onlineSavingsMsg'
Learn More

CD2 Mouse anti-Baboon, Chimpanzee, Cynomolgus Monkey, Human, Non-human primate, Porcine, Rhesus Monkey, FITC, Clone: RPA-2.10, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-0029-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15576296

  • £52.00 / 25 tests

Estimated Shipment date
from Supplier 06-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CD2 (LFA-2) is a monomeric surface antigen (MW range 45-58 kDa) of the human T-lymphocyte lineage that is expressed on all peripheral blood T cells. CD2 is one of the earliest T-cell markers, being present on more than 95% of thymocytes and it is also found on some natural killer cells, but not on B lymphocytes. Monoclonal antibodies directed against CD2 inhibit the formation of rosettes with sheep erythrocytes, indicating that CD2 is the erythrocyte receptor or is closely associated with it. The interaction betwee CD2 and CD58 stabilizes adhesion between T cells and antigen presenting or target cells. Relatively low affinity of CD2 to CD58 (as measured in solution) is compensated within the two-dimensional cell-cell interface to provide tight adhesion. Moreover, T cell activation induces increased CD2 expression and its lateral mobility, making easier contact between CD2 and CD58. Subsequently, T cell activation causes fixation of CD58-CD2 at sites of cell-cell contact, thereby strengthening intercellular adhesion. CD2 deficiency reduces intestinal inflammation and helps to control infection. Diseases associated with CD2 dysfunction include penis squamous cell carcinoma and immune deficiency due to the absence of the thymus.
Specifications

Specifications

CD2
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P06729
CD2 antigen, sheep red blood cell receptor, LFA-3 receptor, T-cell surface antigen CD2, T-cell surface antigen T11/Leu-5, erythrocyte receptor, lymphocyte-function antigen-2, rosette receptor
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Baboon, Chimpanzee, Cynomolgus Monkey, Human, Non-human primate, Porcine, Rhesus Monkey
Flow Cytometry
RPA-2.10
FITC
CD2
Liquid
CD2
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
102120798, 396662, 449512, 677855, 914
Videos
Documents

For Research Use Only.