missing translation for 'onlineSavingsMsg'
Learn More

CD2 Mouse anti-Baboon, Chimpanzee, Cynomolgus Monkey, Human, Non-human primate, Porcine, Rhesus Monkey, APC, Clone: RPA-2.10, eBioscience™

Mouse Monoclonal Antibody

£78.50 - £177.00

Specifications

Antigen CD2
Clone RPA-2.10
Target Species Baboon, Chimpanzee, Cynomolgus Monkey, Human, Non-human primate, Porcine, Rhesus Monkey
Conjugate APC
Isotype IgG1, kappa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15587846 View Documents Affymetrix eBioscience
17-0029-41
25 Tests £78.50
25 tests View alternative products
Save Up to 
»
EU Stock 3 Please to purchase this item. Need a web account? Register with us today!
15597846 View Documents Affymetrix eBioscience
17-0029-42
100 Tests £177.00
100 tests View alternative products
Save Up to 
»
EU Stock 16 Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD2 (LFA-2) is a monomeric surface antigen (MW range 45-58 kDa) of the human T-lymphocyte lineage that is expressed on all peripheral blood T cells. CD2 is one of the earliest T-cell markers, being present on more than 95% of thymocytes and it is also found on some natural killer cells, but not on B lymphocytes. Monoclonal antibodies directed against CD2 inhibit the formation of rosettes with sheep erythrocytes, indicating that CD2 is the erythrocyte receptor or is closely associated with it. The interaction betwee CD2 and CD58 stabilizes adhesion between T cells and antigen presenting or target cells. Relatively low affinity of CD2 to CD58 (as measured in solution) is compensated within the two-dimensional cell-cell interface to provide tight adhesion. Moreover, T cell activation induces increased CD2 expression and its lateral mobility, making easier contact between CD2 and CD58. Subsequently, T cell activation causes fixation of CD58-CD2 at sites of cell-cell contact, thereby strengthening intercellular adhesion. CD2 deficiency reduces intestinal inflammation and helps to control infection. Diseases associated with CD2 dysfunction include penis squamous cell carcinoma and immune deficiency due to the absence of the thymus.
Specifications

Specifications

CD2
Baboon, Chimpanzee, Cynomolgus Monkey, Human, Non-human primate, Porcine, Rhesus Monkey
IgG1, kappa
Monoclonal
Liquid
Antibody
4° C, store in dark, DO NOT FREEZE!
RPA-2.10
APC
Flow Cytometry
Primary
Monoclonal
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
RUO

For Research Use Only.