missing translation for 'onlineSavingsMsg'
Learn More

CD1d, Functional Grade, clone: 1B1, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 16-0011-85

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15278327

  • £187.40 / 500µg


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

Description: The 1B1 monoclonal antibody reacts with mouse CD1d, a 48kDa glycoprotein with structural homology to MHC class I molecules. While similar to MHC Class I, CD1d associates with beta2m, functionally CD1d is similar to MHC Class II. 1B1 detects CD1d at varying levels on mouse leukocytes. 1B1 detects beta2m associated CD1d.Applications Reported: The 1B1 antibody has been reported for use in flow cytometric analysis. 1B1 has also been reported in in vivo and in vitro studies.Applications Tested: The 1B1 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Endotoxin: Less than 0.001 ng/ug antibody as determined by the LAL assay.Storage and handling: Use in a sterile environment.Filtration: 0.2μm post-manufacturing filtered.Purity: Greater than 90%, as determined by SDS-PAGE.Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay.Aggregation: Less than 10%, as determined by HPLC.
Specifications

Specifications

CD1d
Monoclonal
1mg/mL
PBS with no preservative; pH 7.2
P11609
AI747460, CD1.1, Cd1a, Cd1d, Ly-38, T-cell surface glycoprotein CD1d1, antigen-presenting glycoprotein CD1d1
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Functional Assay
1B1
Functional Grade
Cd1d1
Purified
Cd1d1
IgG2b, kappa
500μg
Store at 2-8°C.
Primary
12479
Videos

For Research Use Only.