missing translation for 'onlineSavingsMsg'
Learn More

CD1d, Alexa Fluor 488, clone: 1B1, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 53-0011-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15361970

  • £74.49 / 25µg


Estimated Shipment date
from Supplier 26-03-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The 1B1 monoclonal antibody reacts with mouse CD1d, a 48kDa glycoprotein with structural homology to MHC class I molecules. While similar to MHC Class I, CD1d associates with beta2m, functionally CD1d is similar to MHC Class II. 1B1 detects CD1d at varying levels on mouse leukocytes. 1B1 detects beta2m associated CD1d.Applications Reported: This 1B1 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 1B1 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD1d
1B1
Alexa Fluor 488
Conjugated
Rat
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
CD1.1, Ly-38
IgG2b
RUO
Primary
Murine
Videos