missing translation for 'onlineSavingsMsg'
Learn More

CD1d, clone: 51.1, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-0016-80

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15517406

  • £31.01 / 25µg


Estimated Shipment date
from Supplier 31-05-2018
Add to basket

Description

Description

Description: The monoclonal antibody 51.1 reacts with human CD1d, a member of the CD1 family with similarity to the non-polymorphic MHC Class I-like molecules. CD1d is a highly conserved single transmembrane receptor of the Immunoglobulin Superfamily. CD1d can associate with beta-microglobulin another feature showing similarity to MHC class I molecules, but can also exist as a nonglycosylated protein not in association with beta microglobulin. This suggests different control mechanisms for presenting glycolipid containing molecules to CD1d reactive NKT cells. Expression of CD1d is found on B cells of the periphery, in resting monocytes and cortical thymocytes. On intestinal epithelial cells (IEC) expression is polarized. Expression can also be found at low levels intracellularly in hepatocytes. In HCV (hepatitis C virus) livers, CD1d is highly expressed compared to normal controls.The 51.1 monoclonal antibody has been shown to have functional activity; blocking the interaction of CD1d transfected cells with NKT cells.Applications Reported: This 51.1 antibody has been reported for use in flow cytometric analysis, immunoprecipitation, and immunohistology staining of frozen tissue sections.Applications Tested: This 51.1 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD1d
Monoclonal
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
P15813
AI747460, CD1.1, Cd1a, Cd1d, Ly-38, T-cell surface glycoprotein CD1d1, antigen-presenting glycoprotein CD1d1
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Immunoprecipitation
51.1
Unconjugated
CD1D
Purified
CD1D
IgG2b, kappa
25μg
Store at 2-8°C.
Primary
912
Videos

For Research Use Only.