missing translation for 'onlineSavingsMsg'
Learn More

CD1c Mouse anti-Human, PE, Clone: L161, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-0015-41

Code : NEW

Additional Details : Weight : 0.23750kg

Product Code. 15576666

  • £66.25 / 25 tests

Estimated Shipment
03-11-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the CD1 family of transmembrane glycoproteins, which are structurally related to the major histocompatibility complex (MHC) proteins and form heterodimers with beta-2-microglobulin. The CD1 proteins mediate the presentation of primarily lipid and glycolipid antigens of self or microbial origin to T cells. The human genome contains five CD1 family genes organized in a cluster on chromosome 1. The CD1 family members are thought to differ in their cellular localization and specificity for particular lipid ligands. The protein encoded by this gene localizes to late endosomes and lysosomes via a tyrosine-based motif in the cytoplasmic tail, and requires vesicular acidification to bind lipid antigens.
Specifications

Specifications

CD1c
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P29017
RP11-101J8.3, BDCA1, CD1, CD1A, R7
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
L161
PE
CD1C
Liquid
CD1C
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
911
Videos
Documents

For Research Use Only.