missing translation for 'onlineSavingsMsg'
Learn More

CD1a Mouse anti-Human, PE, Clone: HI149, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-0019-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15536676

  • £60.53 / 25 tests


EU Stock 1
Add to basket

Disclaimers:

For Research Use Only.


Specifications

Description

The HI149 monoclonal antibody reacts with human CD1a, a 49kDa protein expressed by cortical thymocytes and dendritic cells including Langerhans cells. The CD1 family of proteins share some structural and functional characteristics with the MHC class I molecules; however, members of the CD1 family are not polymorphic. Similar to MHC class I, CD1a associates with the beta2-microglobulin and is thought to play a role in antigen presentation.
Description & Specifications

Specifications

CD1a
HI149
PE
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Murine
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (0.5μg)/test
Conjugated
Leu-6, T6, R4
IgG1
RUO
Primary
Human