missing translation for 'onlineSavingsMsg'
Learn More

CD1a, eFluor 450, clone: HI149, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 48-0019-41

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15218867

  • £74.43 / 25 tests


EU Stock 2
Add to basket

Description

Description

Description: The HI149 monoclonal antibody reacts with human CD1a, a 49kDa protein expressed by cortical thymocytes and dendritic cells including Langerhans cells. The CD1 family of proteins share some structural and functional characteristics with the MHC class I molecules; however, members of the CD1 family are not polymorphic. Similar to MHC class I, CD1a associates with the beta2-microglobulin and is thought to play a role in antigen presentation.Applications Reported: This HI149 antibody has been reported for use in flow cytometric analysis.Applications Tested: This HI149 antibody has been pre-titrated and tested by flow cytometric analysis of MOLT-4 cells. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor® 450 is an alternative to Pacific Blue®. eFluor® 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD1a
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P06126
CD1, FCB6, HTA1, R4, T6, CD1A antigen, a polypeptide, T-cell surface antigen T6/Leu-6, T-cell surface glycoprotein CD1a, cluster of differentiation 1 A, cortical thymocyte antigen CD1A
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
HI149
eFluor 450
CD1A
Conjugated
CD1A
IgG1, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
909
Videos

For Research Use Only.