Learn More
CD1a Monoclonal Antibody (HI149), eFluor™ 450, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£220.00
Specifications
Antigen | CD1a |
---|---|
Clone | HI149 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15228867
|
Affymetrix eBioscience
48-0019-42 |
100 Tests |
£220.00
100 tests |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The HI149 monoclonal antibody reacts with human CD1a, a 49 kDa protein expressed by cortical thymocytes and dendritic cells including Langerhans cells. The CD1 family of proteins share some structural and functional characteristics with the MHC class I molecules; however, members of the CD1 family are not polymorphic. Similar to MHC class I, CD1a associates with the beta2-microglobulin and is thought to play a role in antigen presentation. Applications Reported: This HI149 antibody has been reported for use in flow cytometric analysis. Applications Tested: This HI149 antibody has been pre-titrated and tested by flow cytometric analysis of MOLT-4 cells. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor™ 450 is an alternative to Pacific Blue™. eFluor™ 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD1a Monoclonal antibody specifically detects CD1a in Human samples. It is validated for Flow Cytometry.Specifications
CD1a | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P06126 | |
CD1A | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
CD1A |
HI149 | |
Flow Cytometry | |
eFluor 450 | |
Mouse | |
Human | |
909 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.