missing translation for 'onlineSavingsMsg'
Learn More

CD199 (CCR9), FITC, clone: eBioCW-1.2 (CW-1.2), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-1991-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15268189

  • £56.31 / 25µg


EU Stock 4
Add to basket

Description

Description

Description: The eBioCW-1.2 monoclonal antibody reacts with mouse CCR9 (CD199), which is the receptor for thymus-expressed chemokine (TECK). CCR9 is a member of the G protein coupled receptor (GPCR) supergene family, and is involved in trafficking of T cell progenitors within the thymus. CCR9 expression during thymocyte development commences at the double-negative (DN) 3 stage (CD4-CD8-CD25+CD44-), peaks in the double-positive (DP) stage (CD4+CD8+CD25-CD44-), and is down-regulated in commited CD4+ or CD8+ single-positive (SP) thymoyctes. CCR9-deficient mice show a mild impairment in thymoycte development. In the periphery, CCR9 is thought to be expressed on naive CD8+ T cells, but not on naive CD4+ T cells.Applications Reported: This eBioCW-1.2 (CW-1.2) antibody has been reported for use in flow cytometric analysis.Applications Tested: This eBioCW-1.2 (CW-1.2) antibody has been tested by flow cytometric analysis of mouse thymocytes and/or splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD199
Monoclonal
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q9WUT7
CDw199, GPR-9-6, GPR28
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
eBioCW-1.2 (CW-1.2)
FITC
Ccr9
Conjugated
Ccr9
IgG2a
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
12769
Videos

For Research Use Only.