missing translation for 'onlineSavingsMsg'
Learn More

CD199 (CCR9), eFluor 660, clone: BBC3M4, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 50-1999-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15559916

  • £81.61 / 25 tests


EU Stock 1
Add to basket

Description

Description

Description: This BBC3M4 monoclonal antibody reacts with human CD199 (CCR9), a 7 transmembrane spanning G-protein-coupled receptor. CCR9 expression is regulated during thymic development and CCR9 deficiency results in mild impairment in thymocyte development. In the periphery, CCR9 and alpha 4 beta 7 integrin are upregulated in response to dendritic cell-derived retinoic acid, conferring preferential trafficking to gut-associated tissues. CCR9 is the receptor for CCL25 (TECK).Applications Reported: This BBC3M4 antibody has been reported for use in flow cytometric analysis.Applications Tested: This BBC3M4 antibody has been pre-titrated and tested by flow cytometric analysis of stimulated normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD199 (CCR9)
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P51686
CDw199, GPR-9-6, GPR28
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
BBC3M4
eFluor 660
CCR9
Conjugated
CCR9
IgG1, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
10803
Videos

For Research Use Only.