missing translation for 'onlineSavingsMsg'
Learn More

CD197 (CCR7), APC-eFluor 780, clone: 3D12, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 47-1979-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15539576

  • £95.28 / 25 tests


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

Description: The 3D12 monoclonal antibody reacts with human CCR7, also known as EBI-1 and CD197. CCR7 is a member of the G-protein-coupled chemokine receptor family with seven membrane-spanning domains and functions as a receptor for 6Ckine/SLC (secondary lymphoid-tissue chemokine), CCL19 and CCL21. CCR7 has been shown to be internalized via clathrin-coated pits and the majority recycled back to the plasma membrane. CCR7 is expressed on T cells and can be used to distinguish populations of naive from central and effector memory T cells. CCR7 has been shown to play a role in migration of memory T cells to inflamed tissue. Expression of CCR7 is also found on DC's. During DC maturation CCR7 expression increases and is thought to be involved in a variety of functions: chemotaxis to the lymph node, cellular architecture, rate of endocytosis, survival and maturation. Expression of CCR7 on the cell surface can be down regulated upon ligand binding.Applications Reported: This 3D12 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 3D12 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (1 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.It is recommended that the staining incubation time be increased to at least 45 minutes at 2-8°C.APC-eFluor® emits at 780 nm and is excited with the Red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome.Light sensitivity: This tandem is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 633-647 nm; Emission: 780 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD197
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P32248
Bukitt's lymphoma receptor 2, C-C chemokine receptor type 7, CC chemokine receptor 7, CCR-7, CD197, Chemokine (C-C Motif) Receptor 7, chemokine (C-C) receptor 7, CMKBR7, EBI1, EBV-induced G protein-coupled receptor 1, EBV-induced G-protein coupled receptor 1, Epstein-Barr virus induced gene 1, Epstein-Barr Virus-Induced G-Protein Coupled Receptor 1, lymphocyte-specific G protein-coupled peptide receptor, MIP-3 Beta Receptor
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
3D12
APC-eFluor 780
CCR7
Conjugated
CCR7
IgG2a, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage, handling and experimental procedures.
Primary
1236
Videos

For Research Use Only.