missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD196 (CCR6) Monoclonal Antibody (R6H1), PE-Cyanine7, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£105.00 - £240.00
Specifications
Clone | R6H1 |
---|---|
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE-Cyanine7 |
Host Species | Mouse |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12749716
|
Affymetrix eBioscience
25-1969-42 |
100 Tests |
£240.00
Each |
Estimated Shipment: 09-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15588506
|
Affymetrix eBioscience
25-1969-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
CD196 (CCR6) Monoclonal antibody specifically detects CD196 (CCR6) in Human samples. It is validated for Flow Cytometry.Specifications
R6H1 | |
Monoclonal | |
Mouse | |
Human |
Flow Cytometry | |
PE-Cyanine7 | |
RUO | |
Primary |
For Research Use Only.